Product Details

SNP ID
rs140540093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10280280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGATTCGATGTACATGTCAAAA[A/C]TTAAATTATTTCATGCATTACGTCC
Phenotype
MIM: 613379
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CMBL PubMed Links

Gene Details

Gene
CMBL
Gene Name
carboxymethylenebutenolidase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138809.3 1362 UTR 3 NP_620164.1
XM_017009034.1 1362 UTR 3 XP_016864523.1
XM_017009035.1 1362 UTR 3 XP_016864524.1

View Full Product Details