Product Details

SNP ID
rs141106898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:162068051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGCTCAGTCTACTCGACTCCT[C/G]TATTTTCACAGAAAATGACGGTGTG
Phenotype
MIM: 137164
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GABRG2 PubMed Links

Gene Details

Gene
GABRG2
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000816.3 410 Missense Mutation CTA,GTA L18V NP_000807.2
NM_198903.2 410 Missense Mutation CTA,GTA L18V NP_944493.2
NM_198904.2 410 Missense Mutation CTA,GTA L18V NP_944494.1

View Full Product Details