Product Details
- SNP ID
-
rs141261806
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:132419911 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGAAAATGGACTCCCTGGCTGCT[A/G]GAGAGTTGAATGCCAGCCACCAGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C5orf56
PubMed Links
Gene Details
- Gene
- C5orf56
- Gene Name
- chromosome 5 open reading frame 56
There are no transcripts associated with this gene.
View Full Product Details