Product Details

SNP ID
rs141306752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132542028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTAACTTACTTTTTGGCCGTCA[A/C]TGTATTTCTTTATTAAGGACAAGTT
Phenotype
MIM: 147850
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL5 PubMed Links

Gene Details

Gene
IL5
Gene Name
interleukin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000879.2 337 Missense Mutation AGT,ATT S98I NP_000870.1
XM_005271988.3 337 Missense Mutation AGT,ATT S120I XP_005272045.1
XM_011543373.2 337 Missense Mutation AGT,ATT S98I XP_011541675.1
XM_011543374.2 337 Missense Mutation AGT,ATT S98I XP_011541676.1

View Full Product Details