Product Details

SNP ID
rs141336392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134738653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGGCCGTCGCTACCGACGGCGG[A/G]GAGAGGCCGGGGGTCCCAGCGGGCT
Phenotype
MIM: 601118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMLG PubMed Links

Gene Details

Gene
CAMLG
Gene Name
calcium modulating ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001745.3 727 Silent Mutation GGA,GGG G11G NP_001736.1
XM_011543653.1 727 Silent Mutation GGA,GGG G11G XP_011541955.1

View Full Product Details