Product Details

SNP ID
rs141792392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134737971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTAGGCTGACCTACTTACTGCC[A/C]TCCCTTTCCCACTCCCGGTTTTCTT
Phenotype
MIM: 601118
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAMLG PubMed Links

Gene Details

Gene
CAMLG
Gene Name
calcium modulating ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001745.3 45 Intron NP_001736.1
XM_011543653.1 45 UTR 5 XP_011541955.1

View Full Product Details