Product Details

SNP ID
rs142067236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:116444395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCACTTTTTCACTATCCAACTC[A/G]AAACTGTCATTGTCAGTCTTTTTTT
Phenotype
MIM: 616704 MIM: 605885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMMD10 PubMed Links

Gene Details

Gene
COMMD10
Gene Name
COMM domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308080.1 6091 Intron NP_001295009.1
NM_016144.3 6091 Intron NP_057228.1
XM_011543458.2 6091 Intron XP_011541760.1
XM_011543460.2 6091 Intron XP_011541762.1
XM_017009553.1 6091 Intron XP_016865042.1
XM_017009555.1 6091 Intron XP_016865044.1
XM_017009556.1 6091 Intron XP_016865045.1
XM_017009557.1 6091 Intron XP_016865046.1
Gene
SEMA6A
Gene Name
semaphorin 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300780.1 6091 UTR 3 NP_001287709.1
NM_020796.4 6091 UTR 3 NP_065847.1
XM_005272042.3 6091 Intron XP_005272099.2
XM_006714663.1 6091 UTR 3 XP_006714726.1
XM_017009675.1 6091 UTR 3 XP_016865164.1
XM_017009676.1 6091 UTR 3 XP_016865165.1
Gene
SEMA6A-AS1
Gene Name
SEMA6A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details