Product Details
- SNP ID
-
rs142070600
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:151183556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTTCTCCTGCTGGAGCTGTCGCC[A/G]CAGCTGCACCTCCTTCTCCAGGGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC69
PubMed Links
Gene Details
- Gene
- CCDC69
- Gene Name
- coiled-coil domain containing 69
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015621.2 |
895 |
Missense Mutation |
CGG,TGG |
R258W |
NP_056436.2 |
View Full Product Details