Product Details

SNP ID
rs142116239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138951255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTCCTGCACCAGGGTCCTCAG[G/T]ACCTGCAGCCCCCCGAGCTTCAGGA
Phenotype
MIM: 608005
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIL1 PubMed Links

Gene Details

Gene
SIL1
Gene Name
SIL1 nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037633.1 1188 Silent Mutation GTA,GTC V315V NP_001032722.1
NM_022464.4 1188 Silent Mutation GTA,GTC V315V NP_071909.1
XM_011543570.2 1188 Silent Mutation GTA,GTC V325V XP_011541872.1

View Full Product Details