Product Details

SNP ID
rs142254256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:171387987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACATGGACATGAGCCCCCTGAG[A/G]CCCCAGAACTATCTTTTCGGTAACT
Phenotype
MIM: 164040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3912 PubMed Links

Gene Details

Gene
MIR3912
Gene Name
microRNA 3912
There are no transcripts associated with this gene.

Gene
NPM1
Gene Name
nucleophosmin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037738.2 Intron NP_001032827.1
NM_002520.6 Intron NP_002511.1
NM_199185.3 Intron NP_954654.1
XM_005265920.3 Intron XP_005265977.1
XM_011534564.1 Intron XP_011532866.1

View Full Product Details