Product Details

SNP ID
rs142342609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180590643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACGGCCCCCACGGCCTGGGGA[C/T]CCAGCTCAGCCACACACTTCTGGGA
Phenotype
MIM: 606500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCGB3A1 PubMed Links

Gene Details

Gene
SCGB3A1
Gene Name
secretoglobin family 3A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052863.2 273 Missense Mutation GAT,GGT D83G NP_443095.2

View Full Product Details