Product Details

SNP ID
rs142865560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132541904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTCTCCGTCTTTCTTCTCCACA[C/T]TTTTTCTGTGAAAAAAGAAAAATGA
Phenotype
MIM: 147850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL5 PubMed Links

Gene Details

Gene
IL5
Gene Name
interleukin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000879.2 356 Silent Mutation NP_000870.1
XM_005271988.3 356 Missense Mutation XP_005272045.1
XM_011543373.2 356 Missense Mutation XP_011541675.1
XM_011543374.2 356 Missense Mutation XP_011541676.1

View Full Product Details