Product Details

SNP ID
rs143340807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150007499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGTAGAGCAACAGCAACAACTA[C/T]GTTATAGCAGAACAGGTTGTATTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGXB3 PubMed Links

Gene Details

Gene
HMGXB3
Gene Name
HMG-box containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014983.2 Intron NP_055798.2
XM_006714765.2 Intron XP_006714828.1
Gene
TIGD6
Gene Name
tigger transposable element derived 6
There are no transcripts associated with this gene.

View Full Product Details