Product Details

SNP ID
rs143564828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134741200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGACCAGCAGGGTGGTGTGGCC[A/G]AGGTAAAGGGGACCCAACTGGGAGA
Phenotype
MIM: 601118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMLG PubMed Links

Gene Details

Gene
CAMLG
Gene Name
calcium modulating ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001745.3 1004 Missense Mutation AAG,GAG K104E NP_001736.1
XM_011543653.1 1004 Missense Mutation AAG,GAG K104E XP_011541955.1

View Full Product Details