Product Details

SNP ID
rs143700337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177994192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTACTGGTTCCTCCCAAAGGCTG[A/T]CTCCAGCTGTTCCAACTGCACTGGG
Phenotype
MIM: 601538
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PROP1 PubMed Links

Gene Details

Gene
PROP1
Gene Name
PROP paired-like homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006261.4 565 Missense Mutation ACA,TCA T86S NP_006252.3

View Full Product Details