Product Details

SNP ID
rs143745216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163464792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCAGCTAGAAAAGTTAAGTCTT[C/G]GGAATCAAAGGTGAGGAGCTTTTAT
Phenotype
MIM: 600936
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMMR PubMed Links

Gene Details

Gene
HMMR
Gene Name
hyaluronan mediated motility receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142556.1 397 Missense Mutation TCG,TGG S72W NP_001136028.1
NM_001142557.1 397 Intron NP_001136029.1
NM_012484.2 397 Missense Mutation TCG,TGG S72W NP_036616.2
NM_012485.2 397 Missense Mutation TCG,TGG S72W NP_036617.2
Gene
NUDCD2
Gene Name
NudC domain containing 2
There are no transcripts associated with this gene.

View Full Product Details