Product Details

SNP ID
rs144008686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43039921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGTGAGTACTCTCCTAGTAC[G/T]GGATACAAAGGAAGAGGCCACGGCC
Phenotype
MIM: 611296
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANXA2R PubMed Links

Gene Details

Gene
ANXA2R
Gene Name
annexin A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014279.2 425 Silent Mutation CCA,CCC P42P NP_001014301.1
Gene
LOC153684
Gene Name
uncharacterized LOC153684
There are no transcripts associated with this gene.

Gene
LOC648987
Gene Name
uncharacterized LOC648987
There are no transcripts associated with this gene.

View Full Product Details