Product Details

SNP ID
rs144366124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138118841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAGGATCTGCTGGTTTTTGCT[G/T]ACAAAGCTCTGTGAGTCCTTCAAGC
Phenotype
MIM: 603575
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NME5 PubMed Links

Gene Details

Gene
NME5
Gene Name
NME/NM23 family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003551.2 537 Missense Mutation AAG,CAG K178Q NP_003542.1
XM_005272099.1 537 Intron XP_005272156.1
XM_006714716.3 537 Intron XP_006714779.1
XM_017009945.1 537 Missense Mutation AAG,CAG K178Q XP_016865434.1

View Full Product Details