Product Details

SNP ID
rs145027254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69419500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCAACTCTTCATGACAGTGAG[C/T]GGGCAGTGAGCGCTGATCCCTTGCC
Phenotype
MIM: 610572 MIM: 603139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MARVELD2 PubMed Links

Gene Details

Gene
MARVELD2
Gene Name
MARVEL domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038603.2 277 Missense Mutation CGG,TGG R39W NP_001033692.2
NM_001244734.1 277 Missense Mutation CGG,TGG R39W NP_001231663.1
XM_005248445.4 277 Missense Mutation CGG,TGG R39W XP_005248502.1
XM_005248446.4 277 Missense Mutation CGG,TGG R39W XP_005248503.1
XM_005248447.4 277 Missense Mutation CGG,TGG R39W XP_005248504.1
Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
There are no transcripts associated with this gene.

View Full Product Details