Product Details

SNP ID
rs145791175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149376759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTAGCCCCAGGGAGACAGGCT[C/T]CTCTTGTTGGACATCCACAGCTGCA
Phenotype
MIM: 604627
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17B PubMed Links

Gene Details

Gene
IL17B
Gene Name
interleukin 17B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317987.1 661 Silent Mutation AGA,AGG R44R NP_001304916.1
NM_014443.2 661 Silent Mutation AGA,AGG R96R NP_055258.1
XM_011537622.2 661 Silent Mutation AGA,AGG R44R XP_011535924.1
XM_017009346.1 661 Silent Mutation AGA,AGG R161R XP_016864835.1
XM_017009347.1 661 Silent Mutation AGA,AGG R44R XP_016864836.1
XM_017009348.1 661 Silent Mutation AGA,AGG R44R XP_016864837.1
Gene
PCYOX1L
Gene Name
prenylcysteine oxidase 1 like
There are no transcripts associated with this gene.

View Full Product Details