Product Details

SNP ID
rs146789460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177484126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAAGGCCCTGTTCCGGATGGGC[A/G]TCTTGCAGGCAGCACAGGTAAAGCA
Phenotype
MIM: 126660 MIM: 605903
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBN1 PubMed Links

Gene Details

Gene
DBN1
Gene Name
drebrin 1
There are no transcripts associated with this gene.

Gene
PDLIM7
Gene Name
PDZ and LIM domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005451.4 1204 Missense Mutation ACG,ATG T372M NP_005442.2
NM_203352.2 1204 Missense Mutation ACG,ATG T338M NP_976227.1
NM_213636.2 1204 Intron NP_998801.1
XM_011534698.2 1204 Missense Mutation ACG,ATG T279M XP_011533000.1
XM_011534699.2 1204 Missense Mutation ACG,ATG T224M XP_011533001.1

View Full Product Details