Product Details

SNP ID
rs147400420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34936155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGTTTTTTAGGGATTTTACAC[A/G]GTGTATCGTAATGTTTTTGAAATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC21 PubMed Links

Gene Details

Gene
DNAJC21
Gene Name
DnaJ heat shock protein family (Hsp40) member C21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012339.2 554 Silent Mutation ACA,ACG T109T NP_001012339.2
NM_194283.3 554 Silent Mutation ACA,ACG T109T NP_919259.3
XM_005248249.4 554 Silent Mutation ACA,ACG T196T XP_005248306.2
XM_005248250.3 554 Silent Mutation ACA,ACG T196T XP_005248307.2
XM_011513965.2 554 Silent Mutation ACA,ACG T196T XP_011512267.2
XM_011513966.2 554 Silent Mutation ACA,ACG T196T XP_011512268.2

View Full Product Details