Product Details

SNP ID
rs147834789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177303250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCTTGCCCACGTACTCCTTGCC[C/T]AGCATCACCACCTTGACGTCCACGC
Phenotype
MIM: 609450 MIM: 606681 MIM: 605733 MIM: 612415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MXD3 PubMed Links

Gene Details

Gene
MXD3
Gene Name
MAX dimerization protein 3
There are no transcripts associated with this gene.

Gene
NSD1
Gene Name
nuclear receptor binding SET domain protein 1
There are no transcripts associated with this gene.

Gene
PRELID1
Gene Name
PRELI domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271828.1 409 Intron NP_001258757.1
NM_013237.3 409 Intron NP_037369.1
Gene
RAB24
Gene Name
RAB24, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031677.3 409 Silent Mutation CTA,CTG L13L NP_001026847.1
NM_130781.3 409 Silent Mutation CTA,CTG L13L NP_570137.2

View Full Product Details