Product Details

SNP ID
rs147921688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178114924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATGGAGGGCTGGGCAGACATA[C/T]ATGCAATACATACACAGCAGTGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
N4BP3 PubMed Links

Gene Details

Gene
N4BP3
Gene Name
NEDD4 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015111.1 Intron NP_055926.1
XM_006714834.3 Intron XP_006714897.1
XM_011534473.1 Intron XP_011532775.1
XM_011534474.1 Intron XP_011532776.1

View Full Product Details