Product Details

SNP ID
rs148914345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178206871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTGAGGAAAAGATCAGGGAGT[A/T]CTTTGGCGAGTTTGGGGAGGTGAGT
Phenotype
MIM: 602688 MIM: 614683
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPAB PubMed Links

Gene Details

Gene
HNRNPAB
Gene Name
heterogeneous nuclear ribonucleoprotein A/B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004499.3 775 Missense Mutation TAC,TTC Y173F NP_004490.2
NM_031266.2 775 Missense Mutation TAC,TTC Y173F NP_112556.2
Gene
PHYKPL
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278346.1 775 Intron NP_001265275.1
NM_153373.3 775 Intron NP_699204.1

View Full Product Details