Product Details

SNP ID
rs149852902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138558628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGACACACTTTAATTTCCACTTG[C/G]GTTTGACCATCAGCGGCAGTAGAGA
Phenotype
MIM: 600548
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSPA9 PubMed Links

Gene Details

Gene
HSPA9
Gene Name
heat shock protein family A (Hsp70) member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004134.6 1751 Silent Mutation ACC,ACG T480T NP_004125.3
Gene
SNORD63
Gene Name
small nucleolar RNA, C/D box 63
There are no transcripts associated with this gene.

View Full Product Details