Product Details

SNP ID
rs150109655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72107627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGCCTAGCCTGTCGCACCGCTT[C/T]CTTGACAGCAAGTTCTACTTGCTGG
Phenotype
MIM: 157129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1B PubMed Links

Gene Details

Gene
MAP1B
Gene Name
microtubule associated protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324255.1 360 Intron NP_001311184.1
NM_005909.4 360 Silent Mutation TTC,TTT F32F NP_005900.2

View Full Product Details