Product Details

SNP ID
rs150389257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55030470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTCTCCCACTGTGGTTTTAGG[C/T]GCACACTCTCTCTCAAAGAATGAGG
Phenotype
MIM: 600784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GZMK PubMed Links

Gene Details

Gene
GZMK
Gene Name
granzyme K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002104.2 293 Silent Mutation GGC,GGT G83G NP_002095.1
Gene
LOC102467081
Gene Name
uncharacterized LOC102467081
There are no transcripts associated with this gene.

View Full Product Details