Product Details
- SNP ID
-
rs150467271
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:135535611 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTCTGGCACAGTCTTCCTCGTCG[A/G]TGAGGAAGCCGGATAGGTCACTGCC
- Phenotype
-
MIM: 601726
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NEUROG1
PubMed Links
Gene Details
- Gene
- NEUROG1
- Gene Name
- neurogenin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006161.2 |
339 |
Missense Mutation |
ACC,ATC |
T27I |
NP_006152.2 |
View Full Product Details