Product Details

SNP ID
rs41270608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28904259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGTGTGACACCTCTCTGTCAC[A/G]TTGTAGAAGGAGACCTCACCAGCAT
Phenotype
MIM: 602165
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCG14 PubMed Links

Gene Details

Gene
HCG14
Gene Name
HLA complex group 14 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TRIM27
Gene Name
tripartite motif containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006510.4 1712 Silent Mutation AAC,AAT N451N NP_006501.1

View Full Product Details