Product Details
- SNP ID
-
rs138070617
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26385028 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGTTGAACAGCCCAGTTTACTGT[C/T]GTGGGGCCAGCTAATCCCATCCTGG
- Phenotype
-
MIM: 613591
MIM: 613594
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTN2A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61364019] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTN2A2
- Gene Name
- butyrophilin subfamily 2 member A2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001197237.1 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
NP_001184166.1 |
NM_001197238.1 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
NP_001184167.1 |
NM_001197239.1 |
210 |
Intron |
|
|
NP_001184168.1 |
NM_001197240.1 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
NP_001184169.1 |
NM_006995.4 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
NP_008926.2 |
NM_181531.2 |
210 |
Intron |
|
|
NP_853509.1 |
XM_005248797.3 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_005248854.1 |
XM_005248798.4 |
210 |
Intron |
|
|
XP_005248855.1 |
XM_006714953.3 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_006715016.1 |
XM_006714955.3 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_006715018.1 |
XM_011514228.2 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_011512530.1 |
XM_011514229.2 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_011512531.1 |
XM_011514231.2 |
210 |
Intron |
|
|
XP_011512533.1 |
XM_017010166.1 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_016865655.1 |
XM_017010167.1 |
210 |
Intron |
|
|
XP_016865656.1 |
XM_017010168.1 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_016865657.1 |
XM_017010169.1 |
210 |
Silent Mutation |
GTC,GTT |
V36V |
XP_016865658.1 |
- Gene
- BTN3A2
- Gene Name
- butyrophilin subfamily 3 member A2
View Full Product Details