Product Details

SNP ID
rs138864432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:656410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGAGGTTTGCTTCCACCAGCA[C/T]GTGACTGCCCACGTTCGCCATCCTC
Phenotype
MIM: 615329 MIM: 609713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC2 PubMed Links

Gene Details

Gene
EXOC2
Gene Name
exocyst complex component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018303.5 555 Intron NP_060773.3
XM_017011018.1 555 Intron XP_016866507.1
XM_017011019.1 555 Intron XP_016866508.1
XM_017011020.1 555 Intron XP_016866509.1
XM_017011021.1 555 Intron XP_016866510.1
XM_017011022.1 555 Intron XP_016866511.1
XM_017011023.1 555 Intron XP_016866512.1
XM_017011024.1 555 Intron XP_016866513.1
XM_017011025.1 555 Intron XP_016866514.1
XM_017011026.1 555 Intron XP_016866515.1
Gene
HUS1B
Gene Name
HUS1 checkpoint clamp component B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148959.3 555 Missense Mutation NP_683762.2

View Full Product Details