Product Details

SNP ID
rs139052456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31810241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACCTCCAGCCACGAAAGGAGCT[C/T]GTTGCATTTATCCAATATTTTATTT
Phenotype
MIM: 140550 MIM: 140559 MIM: 607282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPA1A PubMed Links

Gene Details

Gene
HSPA1A
Gene Name
heat shock protein family A (Hsp70) member 1A
There are no transcripts associated with this gene.

Gene
HSPA1L
Gene Name
heat shock protein family A (Hsp70) member 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005527.3 1915 Missense Mutation AAG,GAG K578E NP_005518.3
Gene
LSM2
Gene Name
LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated
There are no transcripts associated with this gene.

View Full Product Details