Product Details

SNP ID
rs140876468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32129608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTTGAGTATGCTCTAGAATT[G/T]GGCTGGCTGGATCTGGGCTTACTTC
Phenotype
MIM: 600984
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATF6B PubMed Links

Gene Details

Gene
ATF6B
Gene Name
activating transcription factor 6 beta
There are no transcripts associated with this gene.

Gene
FKBPL
Gene Name
FK506 binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022110.3 443 Missense Mutation CAA,CCA Q58P NP_071393.2

View Full Product Details