Product Details

SNP ID
rs141143854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37171353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAGGTGCTGGAGGCCGTGCGG[C/T]ACTGCCACAACTGCGGGGTGCTCCA
Phenotype
MIM: 164960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIM1 PubMed Links

Gene Details

Gene
PIM1
Gene Name
Pim-1 proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243186.1 899 Missense Mutation CAC,TAC H248Y NP_001230115.1
NM_002648.3 899 Missense Mutation CAC,TAC H157Y NP_002639.1

View Full Product Details