Product Details

SNP ID
rs141215754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26197794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACCTGATGTTAAGCTACATGAA[C/G]ACATTTAATTTCACCACAATATGTC
Phenotype
MIM: 602792 MIM: 602804 MIM: 602811 MIM: 602823 MIM: 602830
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2AD PubMed Links

Gene Details

Gene
HIST1H2AD
Gene Name
histone cluster 1, H2ad
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021065.3 Intron NP_066409.1
Gene
HIST1H2BF
Gene Name
histone cluster 1, H2bf
There are no transcripts associated with this gene.

Gene
HIST1H3D
Gene Name
histone cluster 1, H3d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003530.4 Intron NP_003521.2
Gene
HIST1H4D
Gene Name
histone cluster 1, H4d
There are no transcripts associated with this gene.

Gene
HIST1H4E
Gene Name
histone cluster 1, H4e
There are no transcripts associated with this gene.

View Full Product Details