Product Details

SNP ID
rs141511420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26285290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGGCGTGCTCTGTGTAAGTGAC[A/G]GCGTCACGAATCACGTTCTCCAGGA
Phenotype
MIM: 602828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H4H PubMed Links

Gene Details

Gene
HIST1H4H
Gene Name
histone cluster 1, H4h
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003543.3 Intron NP_003534.1

View Full Product Details