Product Details

SNP ID
rs141653419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31687269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGTGGGGTGCAGTGAGTGGC[C/T]TCAAAGTTGTGCAGATGCTTCCGAG
Phenotype
MIM: 142620 MIM: 610434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD16A PubMed Links

Gene Details

Gene
ABHD16A
Gene Name
abhydrolase domain containing 16A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177515.1 1826 Silent Mutation GAA,GAG E507E NP_001170986.1
NM_021160.2 1826 Silent Mutation GAA,GAG E540E NP_066983.1
Gene
LY6G5C
Gene Name
lymphocyte antigen 6 complex, locus G5C
There are no transcripts associated with this gene.

View Full Product Details