Product Details

SNP ID
rs142160745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37008919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCTCCCGAGTCCCCAGGACCA[C/T]GGGAGAAGACGAATGTCGGGGAGGC
Phenotype
MIM: 605091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGD2 PubMed Links

Gene Details

Gene
FGD2
Gene Name
FYVE, RhoGEF and PH domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173558.3 317 Missense Mutation CGG,TGG R52W NP_775829.2
XM_011514372.2 317 Missense Mutation CGG,TGG R52W XP_011512674.1
XM_011514373.2 317 Missense Mutation CGG,TGG R52W XP_011512675.1
XM_017010429.1 317 Missense Mutation CGG,TGG R52W XP_016865918.1

View Full Product Details