Product Details

SNP ID
rs142640766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152972393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGATTTCTGAACAGAAGCCAGT[A/G]GGGTTCTGAACATAACATATTCTCT
Phenotype
MIM: 606013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO5 PubMed Links

Gene Details

Gene
FBXO5
Gene Name
F-box protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142522.1 1080 Missense Mutation CCA,CTA P278L NP_001135994.1
NM_012177.3 1080 Missense Mutation CCA,CTA P324L NP_036309.1

View Full Product Details