Product Details
- SNP ID
-
rs143216774
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35778387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGATTTCTTGAGAGGAGCATGG[C/T]ACTGCAGTGGCAGGACCACAGCTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLPSL1
PubMed Links
Gene Details
- Gene
- CLPSL1
- Gene Name
- colipase like 1
There are no transcripts associated with this gene.
- Gene
- CLPSL2
- Gene Name
- colipase like 2
View Full Product Details