Product Details

SNP ID
rs143456302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44002084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCCTAGGTGCCTCCAGTTTGAG[A/G]GAGGAGAGAGGACACCCCCTCCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf223 PubMed Links

Gene Details

Gene
C6orf223
Gene Name
chromosome 6 open reading frame 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171992.2 1342 Missense Mutation AGA,GGA R95G NP_001165463.1
NM_001324369.1 1342 UTR 5 NP_001311298.1
NM_153246.5 1342 Silent Mutation AGA,AGG R80R NP_694978.2
XM_017010425.1 1342 Silent Mutation AGA,AGG R79R XP_016865914.1
Gene
LOC101929705
Gene Name
uncharacterized LOC101929705
There are no transcripts associated with this gene.

View Full Product Details