Product Details

SNP ID
rs144070852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:170283311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGTGTCACGCTTGCTGTGCGC[A/G]TCCCTGACGGCGGTGTCGTCACCCT
Phenotype
MIM: 606582 MIM: 612266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLL1 PubMed Links

Gene Details

Gene
DLL1
Gene Name
delta like canonical Notch ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005618.3 2438 Silent Mutation GAC,GAT D656D NP_005609.3
XM_005266934.3 2438 Intron XP_005266991.1
Gene
FAM120B
Gene Name
family with sequence similarity 120B
There are no transcripts associated with this gene.

Gene
LINC01624
Gene Name
long intergenic non-protein coding RNA 1624
There are no transcripts associated with this gene.

Gene
LOC285804
Gene Name
uncharacterized LOC285804
There are no transcripts associated with this gene.

View Full Product Details