Product Details

SNP ID
rs144462878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105278183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACGTTCAGGCACCGCGCGATGAA[C/T]GCAAACATGTCTGAGACTTCCTCTA
Phenotype
MIM: 600400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PREP PubMed Links

Gene Details

Gene
PREP
Gene Name
prolyl endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002726.4 2327 Silent Mutation GCA,GCG A698A NP_002717.3
XM_005267044.2 2327 Intron XP_005267101.1
XM_011535925.2 2327 Intron XP_011534227.1

View Full Product Details