Product Details

SNP ID
rs144867793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:78940777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGTTCCAAAAGTTAAAGAGGT[A/G]TCTTCGAACAACAGCTGCCTTTGCT
Phenotype
MIM: 615375 MIM: 612870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRAK1BP1 PubMed Links

Gene Details

Gene
IRAK1BP1
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010844.3 5348 Intron NP_001010844.1
XM_006715336.3 5348 Intron XP_006715399.1
XM_011535447.1 5348 Intron XP_011533749.1
XM_011535448.2 5348 Intron XP_011533750.1
XM_017010264.1 5348 Intron XP_016865753.1
XM_017010265.1 5348 Intron XP_016865754.1
Gene
PHIP
Gene Name
pleckstrin homology domain interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017934.5 5348 Silent Mutation GAC,GAT D1794D NP_060404.3
XM_005248729.4 5348 Silent Mutation GAC,GAT D1793D XP_005248786.1
XM_011535918.2 5348 Silent Mutation GAC,GAT D1622D XP_011534220.1
XM_011535919.1 5348 Intron XP_011534221.1
XM_017010989.1 5348 Silent Mutation GAC,GAT D1239D XP_016866478.1
XM_017010990.1 5348 Silent Mutation GAC,GAT D1239D XP_016866479.1

View Full Product Details