Product Details

SNP ID
rs145427268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159767084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTCCAGCATGGAGCTGCCAGAT[A/G]ATCTGTGGGTCAGGCCTAAAAGCTG
Phenotype
MIM: 100678 MIM: 147460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAT2 PubMed Links

Gene Details

Gene
ACAT2
Gene Name
acetyl-CoA acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303253.1 401 Missense Mutation ATA,ATG I119M NP_001290182.1
NM_005891.2 401 Missense Mutation ATA,ATG I90M NP_005882.2
Gene
LOC100129518
Gene Name
uncharacterized LOC100129518
There are no transcripts associated with this gene.

Gene
SOD2
Gene Name
superoxide dismutase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details