Product Details

SNP ID
rs145737035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149726906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTCTGATACTAAAATTTTGCA[C/T]AGAAGTTGGATTTCAAAAGGCTAAT
Phenotype
MIM: 603473 MIM: 608141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LATS1 PubMed Links

Gene Details

Gene
LATS1
Gene Name
large tumor suppressor kinase 1
There are no transcripts associated with this gene.

Gene
LOC645967
Gene Name
uncharacterized LOC645967
There are no transcripts associated with this gene.

Gene
NUP43
Gene Name
nucleoporin 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198887.2 1283 UTR 3 NP_942590.1
XM_005266960.4 1283 UTR 3 XP_005267017.1
XM_005266961.3 1283 UTR 3 XP_005267018.1
XM_005266962.3 1283 UTR 3 XP_005267019.1
XM_011535798.2 1283 Intron XP_011534100.1
XM_011535799.2 1283 UTR 3 XP_011534101.1

View Full Product Details