Product Details

SNP ID
rs145805719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28904101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGTCTCCATGGAATGACCATGA[C/T]TCCCAACATGGCCAGAAAACCCATC
Phenotype
MIM: 602165
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCG14 PubMed Links

Gene Details

Gene
HCG14
Gene Name
HLA complex group 14 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TRIM27
Gene Name
tripartite motif containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006510.4 1870 Missense Mutation AAT,AGT N504S NP_006501.1

View Full Product Details