Product Details

SNP ID
rs146384586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132645503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTTGTAATATATCTCTTCAGC[A/G]CCTTTGAAGTTTAGCTCCAGAAAGA
Phenotype
MIM: 609333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAAR1 PubMed Links

Gene Details

Gene
TAAR1
Gene Name
trace amine associated receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138327.2 524 Silent Mutation GGC,GGT G167G NP_612200.1

View Full Product Details