Product Details

SNP ID
rs146483066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89894517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCATCCTAGAGGAAGTTCACTCC[C/T]ACTCAACCATAGTGGGGAAAATCTG
Phenotype
MIM: 611924
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA10 PubMed Links

Gene Details

Gene
GJA10
Gene Name
gap junction protein alpha 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032602.1 49 Missense Mutation CAC,TAC H17Y NP_115991.1

View Full Product Details